missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Pentafluorothio)phenol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Synthesis of hexylamides using fluorine derivatives as intermediate active esters. Various non-metallic derivatives of pentafluorothiophenol, containing sulfur bonded to boron, carbon, silicon, germanium, phosphorus, arsenic, antimony, sulfur, and chlorine, can be prepared.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
Quantity | 5 g |
Formula Weight | 220.16 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Pentafluorothio)phenol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'