Learn More
4-Penten-1-ol, 99%, Thermo Scientific Chemicals
91.56 € - 302.51 €
Chemical Identifiers
CAS | 821-09-0 |
---|---|
Molecular Formula | C5H10O |
Molecular Weight (g/mol) | 86.13 |
MDL Number | MFCD00002975 |
InChI Key | LQAVWYMTUMSFBE-UHFFFAOYSA-N |
Synonym | 4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene |
PubChem CID | 13181 |
IUPAC Name | pent-4-en-1-ol |
SMILES | C=CCCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10665772
|
Thermo Scientific Acros
150010100 |
10 g | Glass bottle |
91.56 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10001630
|
Thermo Scientific Acros
150010500 |
50 g | Glass bottle |
302.51 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
821-09-0 | |
86.13 | |
LQAVWYMTUMSFBE-UHFFFAOYSA-N | |
13181 | |
C=CCCCO |
C5H10O | |
MFCD00002975 | |
4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene | |
pent-4-en-1-ol |
Specifications
821-09-0 | |
100.0 | |
0.8340g/mL | |
43°C | |
98.5% min. (GC) | |
H2C=CH(CH2)3OH | |
MFCD00002975 | |
0.834 | |
Solubility in water: 57g/L (25°C) | |
C=CCCCO | |
86.13 | |
86.13 | |
Liquid |
98.5 | |
Colorless | |
134°C to 137°C | |
Authentic | |
C5H10O | |
1.4289 to 1.4309 | |
01, 443 | |
4-penten-1-ol, 4-pentenol, 4-pentenyl alcohol, 2-allylethyl alcohol, unii-w0558tqc6f, 4-pentene-1-ol, 4-pentenol-1, 4-penten-1ol, 4penten-1-ol, 5-hydroxy-1-pentene | |
LQAVWYMTUMSFBE-UHFFFAOYSA-N | |
pent-4-en-1-ol | |
13181 | |
99% | |
4-Penten-1-ol, 99% |
Safety and Handling
GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 212-473-2
- 4-Penten-1-ol
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only