missing translation for 'onlineSavingsMsg'
Learn More

4-Pentenoic acid, 99%, Thermo Scientific Chemicals

Product Code. 11974961
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-80-0
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00004408
InChI Key HVAMZGADVCBITI-UHFFFAOYSA-N
Synonym 4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1
PubChem CID 61138
ChEBI CHEBI:35936
IUPAC Name pent-4-enoic acid
SMILES OC(=O)CCC=C

Specifications

Melting Point -22.5°C
Color Colorless to Yellow
Density 0.9750g/mL
Boiling Point 83.0°C to 84.0°C (12.0 mmHg)
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2CH2CO2H
Refractive Index 1.4273 to 1.4303
Quantity 100 g
Beilstein 02,425
Specific Gravity 0.975
Solubility Information Solubility in water: insoluble
Formula Weight 100.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Pentenoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'