missing translation for 'onlineSavingsMsg'
Learn More
4-Phenoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 11388606

4-Phenoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11388606
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11388606

Brand: Thermo Scientific Alfa Aesar L17664.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1623-92-3
Molecular Formula C12H9ClO3S
Molecular Weight (g/mol) 268.71
MDL Number MFCD00625748
InChI Key QIZPONOMFWAPRR-UHFFFAOYSA-N
Synonym 4-phenoxybenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-phenoxy, 4-phenoxy-benzenesulfonyl chloride, 4-phenoxyphenylsulfonyl chloride, 4-phenoxybenzenesulphonyl chloride, 4-phenoxy benzene sulfonyl chloride, acmc-1bvtb, chloro 4-phenoxyphenyl sulfone, 4-phenoxybenzene-sulfonyl chloride, 4-phenoxy-benzenesulphonyl chloride
PubChem CID 2794756
IUPAC Name 4-phenoxybenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Melting Point 42°C to 45°C
Boiling Point 180°C (5 mmHg)
Quantity 1 g
UN Number UN3260
Beilstein 2852659
Sensitivity Moisture sensitive
Formula Weight 268.72
Percent Purity 97%
Chemical Name or Material 4-Phenoxybenzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Phenoxybenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.