missing translation for 'onlineSavingsMsg'
Learn More
4-(Phenylazo)diphenylamine, 97%, Thermo Scientific™ Product Code.: 15374978

4-(Phenylazo)diphenylamine, 97%, Thermo Scientific™

Product Code. 15374978
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 15374978

Brand: Thermo Scientific Acros 212011000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 101-75-7
Melting Point 84.0°C to 91.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Molecular Formula C18H15N3
Linear Formula C6H5N=NC6H4NHC6H5
MDL Number MFCD00003023
Quantity 100 g
Beilstein 16,314
Synonym 4-anilinoazobenzene, 4-phenylazo diphenylamine, 4-phenylazodiphenylamine, 4-benzeneazodiphenylamine, benzenamine, n-phenyl-4-phenylazo, azobenzene, 4-anilino, n-phenyl-4-aminoazobenzene, 4-phenylamino azobenzene, diphenylamine, 4-phenylazo, n-phenyl-4-phenylazo aniline
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and ether
InChI Key VXLFYNFOITWQPM-UHFFFAOYSA-N
SMILES N(C1=CC=CC=C1)C1=CC=C(C=C1)N=NC1=CC=CC=C1
IUPAC Name N-phenyl-4-phenyldiazenylaniline
Molecular Weight (g/mol) 273.34
PubChem CID 7575
Formula Weight 273.33
Percent Purity 97%
Chemical Name or Material 4-(Phenylazo)diphenylamine
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Phenylazo)diphenylamine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Phenylazo)diphenylamine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.