missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylazophenol, 97%, Thermo Scientific Chemicals

47.54 € - 132.66 €

Chemical Identifiers

CAS 1689-82-3
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00002330
InChI Key JTSBGMZPPPULTA-UHFFFAOYSA-N
Synonym C.I. 11800, 4-Hydroxyazobenzene, Solvent Yellow 7
PubChem CID 5354198
IUPAC Name 4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES O=C1C=CC(C=C1)=NNC1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10722602
View Documents Promotion Details
Thermo Scientific Acros
417360250
25 g Glass bottle
47.54 €
25g
In Stock
to see stock.
Add to basket
 
10589803
View Documents Promotion Details
Thermo Scientific Acros
417361000
100 g Glass bottle
132.66 €
100g
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

1689-82-3
198.23
JTSBGMZPPPULTA-UHFFFAOYSA-N
5354198
O=C1C=CC(C=C1)=NNC1=CC=CC=C1
C12H10N2O
MFCD00002330
C.I. 11800, 4-Hydroxyazobenzene, Solvent Yellow 7
4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one

Specifications

1689-82-3
100.0
C12H10N2O
16, 96
JTSBGMZPPPULTA-UHFFFAOYSA-N
O=C1C=CC(C=C1)=NNC1=CC=CC=C1
198.23
198.22
Brown
4-Phenylazophenol
96.0
97%
MFCD00002330
C.I. 11800, 4-Hydroxyazobenzene, Solvent Yellow 7
Authentic
4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
5354198
≥96.0%
Fine Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Product Identifier
  • 4-Phenylazophenol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only