missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylazophenol, 98%, Thermo Scientific Chemicals

Product Code. 11490668
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Phenylazophenol is used to varnish, grease, paraffin wax, resin, soap kind of coloring. It is used in printing/solvent based writing inks. It is used for providing transparent metallic effects like gold imitation provided on metalized polyester films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenylazophenol is used to varnish, grease, paraffin wax, resin, soap kind of coloring. It is used in printing/solvent based writing inks. It is used for providing transparent metallic effects like gold imitation provided on metalized polyester films.

Solubility
Soluble in acetone, ethanol, benzene and ether. Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1689-82-3
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.23
MDL Number MFCD00002330
InChI Key JTSBGMZPPPULTA-UHFFFAOYSA-N
Synonym 4-hydroxyazobenzene, 4-phenylazophenol, 4-phenyldiazenyl phenol, solvent yellow 7, p-phenylazophenol, 4-phenylazo phenol, p-hydroxyazobenzene, organol yellow ap, c.i. solvent yellow 7, phenol, 4-phenylazo
PubChem CID 5354198
IUPAC Name 4-(phenylhydrazinylidene)cyclohexa-2,5-dien-1-one
SMILES O=C1C=CC(C=C1)=NNC1=CC=CC=C1

Specifications

Melting Point 152°C to 155°C
Quantity 25 g
Solubility Information Soluble in acetone,ethanol,benzene and ether. Insoluble in water.
Formula Weight 198.23
Percent Purity 98%
Chemical Name or Material 4-Phenylazophenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.