missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylphenol, 97%, Thermo Scientific Chemicals

Product Code. 10011770
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-69-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.21
MDL Number MFCD00002347
InChI Key YXVFYQXJAXKLAK-UHFFFAOYSA-N
Synonym 4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol
PubChem CID 7103
ChEBI CHEBI:34422
IUPAC Name 4-phenylphenol
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point >163.0°C
Color White to Yellow
Boiling Point 307.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5C6H4OH
Quantity 250 g
Beilstein 06,674
Merck Index 15,7416
Solubility Information Solubility in water: 0.7g/L (20°C). Other solubilities: soluble in alkalies,soluble in most common organic solvents
Formula Weight 170.21
Percent Purity 97%
Physical Form Flakes
Chemical Name or Material 4-Phenylphenol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Phenylphenol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.