missing translation for 'onlineSavingsMsg'
Learn More

4-Phenylphenol, 97%, Thermo Scientific Chemicals

Product Code. 11429762
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescence and phosphorescence quantum yields and fluorescence and phosphorescence lifetimes were obtained for 4-phenylphenol adsorbed on filter paper with either NaCl, NaBr, or NaI at 296 and 93 K. The solid-surface fluorescence and phosphorescence quantum yield values and phosphorescence lifetime values were obtained for p-aminobenzoic acid (PABA) and 4-phenylphenol adsorbed on α-cyclodextrin/NaCl mixtures. 4-Phenylphenol is used as an antioxidant and is a potential EDC.

Solubility
Soluble in water (0.7 mg/ml at 20°C), ether, most common organic solvents, methanol, and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-69-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00002347
InChI Key YXVFYQXJAXKLAK-UHFFFAOYSA-N
Synonym 4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol
PubChem CID 7103
ChEBI CHEBI:34422
IUPAC Name 4-phenylphenol
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)O

Specifications

Melting Point 164°C to 168°C
Boiling Point 305°C to 308°C
Flash Point 165°C (329°F)
Quantity 2500 g
UN Number UN3077
Beilstein 1907452
Merck Index 14,7305
Solubility Information Soluble in water (0.7mg/ml at 20°C),ether,most common organic solvents,methanol,and chloroform.
Formula Weight 170.21
Percent Purity 98%
Chemical Name or Material 4-Phenylphenol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.