missing translation for 'onlineSavingsMsg'
Learn More

4-Phenyltetrahydropyran-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10245904
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 182491-21-0
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.24
MDL Number MFCD00085758
InChI Key BWHJLSRDMNLSET-UHFFFAOYSA-N
Synonym 4-phenyltetrahydro-2h-pyran-4-carboxylic acid, 4-phenyl-tetrahydro-pyran-4-carboxylic acid, 4-phenyltetrahydropyran-4-carboxylic acid, 4-phenyl-oxane-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-phenyl, 4-carboxy-4-phenyloxane, 4-carboxy-4-phenyltetrahydro-2h-pyran, 4-carboxytetrahydro-2h-pyran-4-yl benzene, 4-phenyltetrahydro-2h-pyran-4-carboxylicacid, 4-phenyl-tetrahydro-2h-pyran-4-carboxylic acid
PubChem CID 4138553
IUPAC Name 4-phenyloxane-4-carboxylic acid
SMILES OC(=O)C1(CCOCC1)C1=CC=CC=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 4-Phenyltetrahydropyran-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'