missing translation for 'onlineSavingsMsg'
Learn More

4-Picoline N-oxide, 98%, Thermo Scientific Chemicals

Product Code. 11305079
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-67-4
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006210
InChI Key IWYYIZOHWPCALJ-UHFFFAOYSA-N
Synonym 4-picoline-n-oxide, 4-picoline n-oxide, 4-methylpyridine n-oxide, 4-methylpyridine 1-oxide, pyridine, 4-methyl-, 1-oxide, 4-picoline 1-oxide, 4-picoline, 1-oxide, gamma-picoline n-oxide, gamma-picoline 1-oxide, 4-methylpyridin-1-ium-1-olate
PubChem CID 13857
SMILES CC1=CC=[N+]([O-])C=C1

Specifications

Melting Point 182°C to 185°C
Boiling Point 152°C (11 mmHg)
Flash Point 149°C (300°F)
Quantity 25 g
Beilstein 106329
Sensitivity Hygroscopic
IUPAC Name 4-methyl-1-oxidopyridin-1-ium
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 4-Picoline N-oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.