missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilac™ 4 Pivoting Wheel Dolly
4 Pivoting Wheel Dolly benefits from excellent stability and great manoeuvrability thanks to its 4 ball bearing mounted drive wheels.
Brand: Gilac™ G170144
This item is not returnable.
View return policy
Description
- Universal dolly on wheels.
- Excellent stability and great manoeuvrability thanks to its 4 ball bearing mounted drive wheels.
- 250 kg load capacity.
- Outstanding quality, outclassing usual industry standards.
- ABS injected base with solid frame guaranteeing unrivalled durability.
- 100 mm diameter black PP wheels.
- GILAC removable pulling bar available (as an option).
- Compatible with all GILAC 600 x 400 and 400 x 300 mm containers and crates.
- Dolly also available in black ABS-PC (G170144).
- Stainless steel clevis recommended in humid environments (no damage to the bearings even in the presence of water).
Specifications
Dolly | |
551 lb. | |
ABS Plastic, Chrome-plated Steel | |
Black |
Containers and Crates | |
250 kg | |
100 mm | |
Steel Wheel Housing, 2 Brakes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur