missing translation for 'onlineSavingsMsg'
Learn More
4-Propylbenzoic acid, 97%, Thermo Scientific™ Product Code.: 10647831

4-Propylbenzoic acid, 97%, Thermo Scientific™

Product Code. 10647831
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10647831

Brand: Thermo Scientific Acros 305960050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2438-05-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00013996
InChI Key ATZHGRNFEFVDDJ-UHFFFAOYSA-M
ChEBI CHEBI:34620

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 140.0°C to 144.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3CH2CH2C6H4CO2H
Quantity 5 g
Synonym chinomethionate, chinomethionat, quinomethionate, oxythioquinox, morestan, cetactaelate, forstan, morestane, joust, morestan 2
SMILES CCCC1=CC=C(C=C1)C([O-])=O
IUPAC Name 6-methyl-[1,3]dithiolo[4,5-b]quinoxalin-2-one
PubChem CID 17109
Formula Weight 164.2
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 4-Propylbenzoic acid, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Propylbenzoic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.