missing translation for 'onlineSavingsMsg'
Learn More

4-Propylbenzoic acid, 97%, Thermo Scientific™

Product Code. 10745843
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2438-05-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00013996
InChI Key ATZHGRNFEFVDDJ-UHFFFAOYSA-M

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 140.0°C to 144.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3CH2CH2C6H4CO2H
Quantity 1 g
Synonym 4-n-propylbenzoic acid, benzoic acid, 4-propyl, 4-propyl-benzoic acid, p-n-propyl benzoic acid, p-propylbenzoic acid, unii-3p1v85e45o, 4-propylbenzoicacid, p-propyl benzoic acid, 4-propylbenzolic acid, pubchem1348
SMILES CCCC1=CC=C(C=C1)C(=O)O
IUPAC Name 4-propylbenzoic acid
PubChem CID 137601
Formula Weight 164.2
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 4-Propylbenzoic acid, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'