missing translation for 'onlineSavingsMsg'
Learn More
Alfa Aesar™ (4-Pyridyl)acetone, 98% Product Code.: 11389445

Alfa Aesar™ (4-Pyridyl)acetone, 98%

Product Code. 11389445
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11389445

Brand: Alfa Aesar™ L18667.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
(4-Pyridyl)acetone is a reactant in the synthesis of Milrinone (M344680), a selective phosphodiesterase inhibitor with vasodilating and positive inotropic activity. Cardiotonic.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6304-16-1
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD00129043
InChI Key ILRVKOYYFFNXDB-UHFFFAOYSA-N
Synonym 4-pyridyl acetone, 1-4-pyridinyl-2-propanone, 1-pyridin-4-yl propan-2-one, 1-4-pyridyl acetone, 4-acetonylpyridine, 4-pyridinylacetone, 4-pyridylacetone, unii-r6onm9f2au, 2-propanone, 1-4-pyridinyl
PubChem CID 80552
IUPAC Name 1-pyridin-4-ylpropan-2-one
SMILES CC(=O)CC1=CC=NC=C1

Specifications

Melting Point 13°C
Boiling Point 143°C (20 mmHg)
Refractive Index 1.5225
Quantity 1 g
Beilstein 110960
Solubility Information Slightly soluble in water.
Formula Weight 135.17
Percent Purity 98%
Chemical Name or Material (4-Pyridyl)acetone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR16F040 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10139843 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10175308 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Alfa Aesar™ (4-Pyridyl)acetone, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.