missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylcatechol, 99%, Thermo Scientific Chemicals

Product Code. 10163010
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-29-3
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00002201
InChI Key XESZUVZBAMCAEJ-UHFFFAOYSA-N
Synonym 4-tert-butylcatechol, p-tert-butylcatechol, 4-tert-butylpyrocatechol, p-tert-butyl catechol, 4-tert-butyl benzene-1,2-diol, 4-t-butylpyrocatechol, p-tert-butylpyrocatechol, 4-t-butylcatechol, 1,2-benzenediol, 4-1,1-dimethylethyl, 4-tert-butylcatechin
PubChem CID 7381
IUPAC Name 4-tert-butylbenzene-1,2-diol
SMILES CC(C)(C)C1=CC=C(O)C(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53.0°C to 56.0°C
Density 1.0490g/mL
Boiling Point 285.0°C
Flash Point 129°C
Assay Percent Range 0.7% max. Catechol (GC)
Packaging Plastic bottle
Linear Formula (CH3)3CC6H3(OH)2
Quantity 100 g
Beilstein 06,IV,6014
Fieser 15,54
Specific Gravity 1.049
Solubility Information Solubility in water: 0.2% (25°C). Other solubilities: soluble in hot water,very sol. in alcohol,ethers,esters and ketones,partly sol. in chlorinated and aromatic hydrocarb.,slightly sol. in aliphatics. 240g/100g acetone,80g/100g benzene;60g/100g
Viscosity 90 mm2/s (25°C)
Formula Weight 166.22
Percent Purity 99%
Grade Technical
Chemical Name or Material 4-tert-Butylcatechol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butylcatechol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-(1,1-dimethylethyl)-1,2-benzenediol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.