missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-tert-Butyldimethylsiloxy-1-butanol, 97%, Thermo Scientific Chemicals
48.24 € - 182.00 €
Chemical Identifiers
CAS | 87184-99-4 |
---|---|
Molecular Formula | C10H24O2Si |
Molecular Weight (g/mol) | 204.39 |
MDL Number | MFCD04974070 |
InChI Key | IJEMXJANZPVITP-UHFFFAOYSA-N |
Synonym | 4-tert-butyldimethylsilyl oxy butan-1-ol, 4-dimethyl-tert-butylsilyl oxybutan-1-ol, 4-dimethyl-tert-butylsilyloxy butan-1-ol, 4-tert-butyldimethylsilyl oxy-1-butanol, 1-butanol, 4-1,1-dimethylethyl dimethylsilyl oxy, acmc-20amup, 4-t-butyldimethylsiloxy-1-butanol, 4-t-butyldimethylsilyloxy-1-butanol, 4-tert-butyldimethylsiloxy-1-butanol |
PubChem CID | 186403 |
IUPAC Name | 4-[tert-butyl(dimethyl)silyl]oxybutan-1-ol |
SMILES | CC(C)(C)[Si](C)(C)OCCCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15421498
|
Thermo Scientific Alfa Aesar
H55285.06 |
5 g |
48.24 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15431498
|
Thermo Scientific Alfa Aesar
H55285.14 |
25 g |
182.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySlightly Soluble in water.(2.0 g/L) (25°C). Soluble in Dichloromethane, Ethyl Acetate, Methanol
Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
Chemical Identifiers
87184-99-4 | |
204.39 | |
IJEMXJANZPVITP-UHFFFAOYSA-N | |
186403 | |
CC(C)(C)[Si](C)(C)OCCCCO |
C10H24O2Si | |
MFCD04974070 | |
4-tert-butyldimethylsilyl oxy butan-1-ol, 4-dimethyl-tert-butylsilyl oxybutan-1-ol, 4-dimethyl-tert-butylsilyloxy butan-1-ol, 4-tert-butyldimethylsilyl oxy-1-butanol, 1-butanol, 4-1,1-dimethylethyl dimethylsilyl oxy, acmc-20amup, 4-t-butyldimethylsiloxy-1-butanol, 4-t-butyldimethylsilyloxy-1-butanol, 4-tert-butyldimethylsiloxy-1-butanol | |
4-[tert-butyl(dimethyl)silyl]oxybutan-1-ol |
Specifications
63°C to 66°C (0.3mmHg) | |
C10H24O2Si | |
MFCD04974070 | |
4-tert-butyldimethylsilyl oxy butan-1-ol, 4-dimethyl-tert-butylsilyl oxybutan-1-ol, 4-dimethyl-tert-butylsilyloxy butan-1-ol, 4-tert-butyldimethylsilyl oxy-1-butanol, 1-butanol, 4-1,1-dimethylethyl dimethylsilyl oxy, acmc-20amup, 4-t-butyldimethylsiloxy-1-butanol, 4-t-butyldimethylsilyloxy-1-butanol, 4-tert-butyldimethylsiloxy-1-butanol | |
IJEMXJANZPVITP-UHFFFAOYSA-N | |
4-[tert-butyl(dimethyl)silyl]oxybutan-1-ol | |
186403 | |
97% | |
0.888 |
87184-99-4 | |
(CH3)3CSi(CH3)2O(CH2)4OH | |
105°C (221°F) | |
Slightly Soluble in water.(2.0g/L) (25°C). Soluble in Dichloromethane,Ethyl Acetate,Methanol | |
CC(C)(C)[Si](C)(C)OCCCCO | |
204.39 | |
204.38 | |
1.439 | |
4-tert-Butyldimethylsiloxy-1-butanol |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only