Learn More
4-tert-Butyldimethylsiloxy-3-penten-2-one, 95%, mixture of isomers, Thermo Scientific™
56.60 € - 153.00 €
Chemical Identifiers
CAS | 69404-97-3 |
---|---|
Molecular Formula | C11H22O2Si |
Molecular Weight (g/mol) | 214.38 |
MDL Number | MFCD00010655 |
InChI Key | ZRVOCNOYAJIAAP-CSKARUKUSA-N |
Synonym | 2-t-butyldimethylsiloxy pent-2-en-4-one, 3e-4-tert-butyldimethylsilyl oxy pent-3-en-2-one, 4-tert-butyldimethylsiloxy-3-penten-2-one, 2-tert-butyldimethylsiloxy pent-2-en-4-one, 4-tert-butyldimethylsilyl oxy pent-3-en-2-one, acetylacetone enol tert-butyldimethylsilyl ether, 2-tert-butyldimethylsilyl oxy-2-penten-4-one, e-4-tert-butyldimethylsilyl oxy pent-3-en-2-one, e-4-tert-butyl dimethyl silyl oxypent-3-en-2-one, 4-tert-butyldimethylsiloxy-3-penten-2-one gc |
PubChem CID | 6034727 |
IUPAC Name | (E)-4-[tert-butyl(dimethyl)silyl]oxypent-3-en-2-one |
SMILES | CC(=CC(=O)C)O[Si](C)(C)C(C)(C)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
69404-97-3 | |
214.38 | |
ZRVOCNOYAJIAAP-CSKARUKUSA-N | |
6034727 | |
CC(=CC(=O)C)O[Si](C)(C)C(C)(C)C |
C11H22O2Si | |
MFCD00010655 | |
2-t-butyldimethylsiloxy pent-2-en-4-one, 3e-4-tert-butyldimethylsilyl oxy pent-3-en-2-one, 4-tert-butyldimethylsiloxy-3-penten-2-one, 2-tert-butyldimethylsiloxy pent-2-en-4-one, 4-tert-butyldimethylsilyl oxy pent-3-en-2-one, acetylacetone enol tert-butyldimethylsilyl ether, 2-tert-butyldimethylsilyl oxy-2-penten-4-one, e-4-tert-butyldimethylsilyl oxy pent-3-en-2-one, e-4-tert-butyl dimethyl silyl oxypent-3-en-2-one, 4-tert-butyldimethylsiloxy-3-penten-2-one gc | |
(E)-4-[tert-butyl(dimethyl)silyl]oxypent-3-en-2-one |
Specifications
50°C to 51°C (0.2mmHg) | |
C11H22O2Si | |
73°C (163°F) | |
ZRVOCNOYAJIAAP-CSKARUKUSA-N | |
(E)-4-[tert-butyl(dimethyl)silyl]oxypent-3-en-2-one | |
6034727 | |
95% | |
0.898 |
69404-97-3 | |
MFCD00010655 | |
2-t-butyldimethylsiloxy pent-2-en-4-one, 3e-4-tert-butyldimethylsilyl oxy pent-3-en-2-one, 4-tert-butyldimethylsiloxy-3-penten-2-one, 2-tert-butyldimethylsiloxy pent-2-en-4-one, 4-tert-butyldimethylsilyl oxy pent-3-en-2-one, acetylacetone enol tert-butyldimethylsilyl ether, 2-tert-butyldimethylsilyl oxy-2-penten-4-one, e-4-tert-butyldimethylsilyl oxy pent-3-en-2-one, e-4-tert-butyl dimethyl silyl oxypent-3-en-2-one, 4-tert-butyldimethylsiloxy-3-penten-2-one gc | |
CC(=CC(=O)C)O[Si](C)(C)C(C)(C)C | |
214.38 | |
214.38 | |
Moisture sensitive | |
4-tert-Butyldimethylsiloxy-3-penten-2-one, mixture of isomers |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.