missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenol, 97%, Thermo Scientific Chemicals

Product Code. 10637442
Click to view available options
Quantity:
5 g
1 kg
2.5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-54-4
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00002367
InChI Key QHPQWRBYOIRBIT-UHFFFAOYSA-N
Synonym 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
PubChem CID 7393
ChEBI CHEBI:34444
IUPAC Name 4-tert-butylphenol
SMILES CC(C)(C)C1=CC=C(C=C1)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 96.0°C to 100.0°C
Color White to Beige
Density 0.9000g/mL
Boiling Point 236.0°C to 238.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula (CH3)3CC6H4OH
Refractive Index 1.4787
Quantity 1 kg
Beilstein 06, 524
Merck Index 15, 1588
Specific Gravity 0.9
Solubility Information Solubility in water: 0.6g/L (20°C). Other solubilities: soluble in aq. alcoholic alkali,soluble in alcohol and ether
Viscosity 2.4 mPa.s (110°C)
Formula Weight 150.22
Percent Purity 97%
Physical Form Flakes or Pastilles
Chemical Name or Material 4-tert-Butylphenol, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'