missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Butylphenol, 99%, Thermo Scientific Chemicals

Product Code. 11442578
Click to view available options
Quantity:
500 g
2500 g
10 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

4-tert-butylphenol on condensation with formaldehyde gives calix[5]arene which is used in enzyme mimetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-butylphenol on condensation with formaldehyde gives calix[5]arene which is used in enzyme mimetics.

Solubility
Soluble in water. [8.7 g/L (20°C)]

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-54-4
Molecular Formula C10H14O
MDL Number MFCD00002367
InChI Key QHPQWRBYOIRBIT-UHFFFAOYSA-N
Synonym 4-tert-butyl phenol, butylphen, p-tert-butylphenol, ptbp, 4-1,1-dimethylethyl phenol, 4-t-butylphenol, p-t-butyl phenol, phenol, 4-1,1-dimethylethyl, p-t-butylphenol, p-terc.butylfenol
PubChem CID 7393
ChEBI CHEBI:34444
IUPAC Name 4-tert-butylphenol

Specifications

Melting Point 97°C to 101°C
Density 0.908
Boiling Point 236°C to 238°C
Flash Point 113°C (235°F)
Assay Percent Range 99%
Quantity 500 g
UN Number UN3077
Beilstein 1817334
Merck Index 14,1585
Solubility Information Soluble in water. [8.7g/L (20°C)]
SMILES CC(C)(C)C1=CC=C(C=C1)O
Molecular Weight (g/mol) 150.221
Formula Weight 150.22
Percent Purity 99%
Chemical Name or Material 4-tert-Butylphenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-tert-Butylphenol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H361f-Suspected of damaging fertility.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 4-tert-Butylphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.