missing translation for 'onlineSavingsMsg'
Learn More

4-tert-Pentylcyclohexanone, 97%, Thermo Scientific Chemicals

Product Code. 11485223
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It finds its application as a masking agent and a perfuming agent in cosmetic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a masking agent and a perfuming agent in cosmetic industry.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16587-71-6
Molecular Formula C11H20O
Molecular Weight (g/mol) 168.28
MDL Number MFCD00013798
InChI Key DCSKAMGZSIRJAQ-UHFFFAOYSA-N
Synonym 4-tert-pentyl cyclohexanone, 4-tert-amylcyclohexanone, 4-tert-pentylcyclohexanone, cyclohexanone, 4-1,1-dimethylpropyl, 4-2-methylbutan-2-yl cyclohexan-1-one, 4-1,1-dimethylpropyl cyclohexanone, cyclohexanone, 4-tert-pentyl, orivone, unii-5380bwu79x, 4-tert-pentyl-cyclohexanone
PubChem CID 27941
IUPAC Name 4-(2-methylbutan-2-yl)cyclohexan-1-one
SMILES CCC(C)(C)C1CCC(=O)CC1

Specifications

Density 0.92
Boiling Point 124°C to 125°C (16 mmHg)
Refractive Index 1.469
Quantity 50 g
Beilstein 2240475
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 168.28
Percent Purity 97%
Chemical Name or Material 4-tert-Pentylcyclohexanone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'