missing translation for 'onlineSavingsMsg'
Learn More

4-(Tetrahydropyran-4-yloxy)aniline, 97%, Thermo Scientific™

Product Code. 10772394
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 917483-71-7
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD09064968
InChI Key CCIFXNITZHWCRR-UHFFFAOYSA-N
Synonym 4-tetrahydropyran-4-yloxy aniline, 4-oxan-4-yloxy aniline, 4-tetrahydro-2h-pyran-4-yl oxy aniline, 4-oxan-4-yl oxy aniline, 4-tetrahydropyran-4-yloxy phenylamine, 4-tetrahydro-2h-pyran-4-yloxy aniline, 4-tetrahydro-pyran-4-yloxy-phenylamine, 4-2h-3,4,5,6-tetrahydropyran-4-yloxy phenylamine
PubChem CID 24229526
IUPAC Name 4-(oxan-4-yloxy)aniline
SMILES C1COCCC1OC2=CC=C(C=C2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 193.24
Percent Purity 97%
Chemical Name or Material 4-(Tetrahydropyran-4-yloxy)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'