missing translation for 'onlineSavingsMsg'
Learn More

4-Tetrahydropyranyl methanesulfonate, 95%, Thermo Scientific Chemicals

Product Code. 15456927
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134419-59-3
Molecular Formula C6H12O4S
Molecular Weight (g/mol) 180.218
MDL Number MFCD11505063
InChI Key GSEZHCLWHDZJAB-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-yl methanesulfonate, 2h-pyran-4-ol, tetrahydro-, methanesulfonate, acmc-20mvcv, 4-oxanyl methanesulfonate, tetrahydropyran-4-ol methane sulfonate, tetrahydro-2h-pyran-4-ol methanesulfonate, tetrahydro-2h-pyran-4-yl methanesulphonate, methansulfonic acid tetrahydropyran-4-yl ester, methanesulfonic acid tetrahydropyran-4-yl ester
PubChem CID 11344352
IUPAC Name oxan-4-yl methanesulfonate
SMILES CS(=O)(=O)OC1CCOCC1

Specifications

Quantity 250 mg
Formula Weight 180.22
Percent Purity 95%
Chemical Name or Material 4-Tetrahydropyranyl methanesulfonate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'