missing translation for 'onlineSavingsMsg'
Learn More

4-(thien-2-ylmethyl)benzoic acid, Thermo Scientific™

Product Code. 10706925
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1002727-90-3
Molecular Formula C12H10O2S
Molecular Weight (g/mol) 218.27
InChI Key OAIBKSRZDOMJKZ-UHFFFAOYSA-N
Synonym 4-thien-2-ylmethyl benzoic acid, 4-2-thienylmethyl benzoic acid, 4-thiophen-2-ylmethyl benzoic acid, 4-thiophen-2-yl methyl benzoic acid
PubChem CID 20038987

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 218.27
Percent Purity 97%
Chemical Name or Material 4-(thien-2-ylmethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'