missing translation for 'onlineSavingsMsg'
Learn More

4-Thien-2-yltetrahydropyran-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10745425
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 880166-18-7
Molecular Formula C10H12O3S
Molecular Weight (g/mol) 212.26
MDL Number MFCD09817516
InChI Key VWYOZQWRYINSBP-UHFFFAOYSA-N
Synonym 4-thiophen-2-yl oxane-4-carboxylic acid, 4-thien-2-yltetrahydropyran-4-carboxylic acid, 4-thien-2-yltetrahydro-2h-pyran-4-carboxylic acid, 2h-pyran-4-carboxylicacid, tetrahydro-4-2-thienyl, 4-thiophen-2-yl tetrahydro-2h-pyran-4-carboxylic acid, 4-thiophen-2-yl oxan-4-carboxylic acid, 4-2-thienyl tetrahydropyran-4-carboxylic acid, 4-thien-2-yl tetrahydro-2h-pyran-4-carboxylic acid, 4-2-thienyl-2h-3,4,5,6-tetrahydropyran-4-carboxylic acid
PubChem CID 19066254
SMILES OC(=O)C1(CCOCC1)C1=CC=CS1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 4-(thiophen-2-yl)oxane-4-carboxylic acid
Formula Weight 212.27
Percent Purity 97%
Chemical Name or Material 4-thien-2-yltetrahydropyran-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'