missing translation for 'onlineSavingsMsg'
Learn More

(4-Thien-2-yltetrahydropyran-4-yl)methyl toluene-4-sulfonate, 97%, Thermo Scientific™

49.90 € - 119.00 €

Chemical Identifiers

CAS 921938-87-6
Molecular Formula C17H20O4S2
Molecular Weight (g/mol) 352.463
MDL Number MFCD09879933
InChI Key OZWQPINOYPXHPX-UHFFFAOYSA-N
Synonym 4-thien-2-yltetrahydropyran-4-yl methyl toluene-4-sulfonate, 4-thiophen-2-yl oxan-4-yl methyl 4-methylbenzenesulfonate, 4-thien-2-yltetrahydro-2h-pyran-4-yl methyl 4-methylphenylsulphonate, 4-thiophen-2-yl oxan-4-yl methyl 4-methylbenzene-1-sulfonate, 4-thiophen-2-yl tetrahydro-2h-pyran-4-yl methyl 4-methylbenzenesulfonate, 4-thien-2-yltetrahydro-2h-pyran-4-yl methyl 4-methylphenylsulfonate, 4-thiophen-2-yltetrahydropyran-4-yl methyl toluene-4-sulfonate
PubChem CID 24229670
IUPAC Name (4-thiophen-2-yloxan-4-yl)methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2(CCOCC2)C3=CC=CS3
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10462314
View Documents
Thermo Scientific Maybridge
CC62244CB
250 mg Amber Glass Bottle
49.90 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10397494
View Documents
Thermo Scientific Maybridge
CC62244DA
1 g Amber Glass Bottle
119.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

921938-87-6
352.463
OZWQPINOYPXHPX-UHFFFAOYSA-N
24229670
CC1=CC=C(C=C1)S(=O)(=O)OCC2(CCOCC2)C3=CC=CS3
C17H20O4S2
MFCD09879933
4-thien-2-yltetrahydropyran-4-yl methyl toluene-4-sulfonate, 4-thiophen-2-yl oxan-4-yl methyl 4-methylbenzenesulfonate, 4-thien-2-yltetrahydro-2h-pyran-4-yl methyl 4-methylphenylsulphonate, 4-thiophen-2-yl oxan-4-yl methyl 4-methylbenzene-1-sulfonate, 4-thiophen-2-yl tetrahydro-2h-pyran-4-yl methyl 4-methylbenzenesulfonate, 4-thien-2-yltetrahydro-2h-pyran-4-yl methyl 4-methylphenylsulfonate, 4-thiophen-2-yltetrahydropyran-4-yl methyl toluene-4-sulfonate
(4-thiophen-2-yloxan-4-yl)methyl 4-methylbenzenesulfonate

Specifications

921938-87-6
100.0
C17H20O4S2
4-thien-2-yltetrahydropyran-4-yl methyl toluene-4-sulfonate, 4-thiophen-2-yl oxan-4-yl methyl 4-methylbenzenesulfonate, 4-thien-2-yltetrahydro-2h-pyran-4-yl methyl 4-methylphenylsulphonate, 4-thiophen-2-yl oxan-4-yl methyl 4-methylbenzene-1-sulfonate, 4-thiophen-2-yl tetrahydro-2h-pyran-4-yl methyl 4-methylbenzenesulfonate, 4-thien-2-yltetrahydro-2h-pyran-4-yl methyl 4-methylphenylsulfonate, 4-thiophen-2-yltetrahydropyran-4-yl methyl toluene-4-sulfonate
CC1=CC=C(C=C1)S(=O)(=O)OCC2(CCOCC2)C3=CC=CS3
352.463
352.47
(4-Thien-2-yltetrahydropyran-4-yl)methyl toluene-4-sulfonate
96.5
97%
MFCD09879933
OZWQPINOYPXHPX-UHFFFAOYSA-N
(4-thiophen-2-yloxan-4-yl)methyl 4-methylbenzenesulfonate
24229670
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.