missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4-Thien-3-ylphenyl)methanol, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 4-thien-3-ylphenyl methanol, 4-thiophen-3-yl phenyl methanol, benzenemethanol, 4-3-thienyl, 4-3-thienyl phenyl methanol, 4-thien-3-yl-benzyl alcohol, benzenemethanol,4-3-thienyl, 4-3-thienyl-phenyl-methanol, 4-3-thienyl phenyl methan-1-ol |
Formula Weight | 190.27 |
Percent Purity | 97% |
Chemical Name or Material | (4-Thien-3-ylphenyl)methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'