missing translation for 'onlineSavingsMsg'
Learn More
4-Thiouracil, 97%, Thermo Scientific Chemicals Product Code.: 15478508

4-Thiouracil, 97%, Thermo Scientific Chemicals

Product Code. 15478508
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15478508

Brand: Thermo Scientific Alfa Aesar H61919.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-28-6
Molecular Formula C4H4N2OS
Molecular Weight (g/mol) 128.15
MDL Number MFCD00090842
InChI Key OVONXEQGWXGFJD-UHFFFAOYSA-N
Synonym 4-thiouracil, 2-hydroxy-4 1h-pyrimidinethione, 4-thioxo-3,4-dihydropyrimidin-2 1h-one, 2-hydroxy-4-mercaptopyrimidine, uracil, 4-thio, 4-mercaptopyrimidin-2-ol, 4-sulfanylpyrimidin-2-ol, 2-hydroxy-4-thiopyrimidine, 2 1h-pyrimidinone, 3,4-dihydro-4-thioxo, 2 1h-pyrimidinone, 4-mercapto-9ciShow MoreShow Less
PubChem CID 2734394
IUPAC Name 4-sulfanylidene-1,2,3,4-tetrahydropyrimidin-2-one
SMILES O=C1NC=CC(=S)N1

Specifications

Melting Point 295°C
Quantity 1 g
Formula Weight 128.15
Percent Purity 97%
Chemical Name or Material 4-Thiouracil

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ19K014 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberH61919.03
Lot NumberT18I012 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog NumberH61919.03
Lot NumberN13F032 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61919.03
Lot NumberT23E059 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61919.03
Lot NumberY05F125 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61919.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Thiouracil, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.