Learn More
4-(Trifluoromethoxy)aniline, 99%, Thermo Scientific Chemicals
27.50 € - 117.00 €
Chemical Identifiers
CAS | 461-82-5 |
---|---|
Molecular Formula | C7H6F3NO |
Molecular Weight (g/mol) | 177.12 |
MDL Number | MFCD00041314 |
InChI Key | XUJFOSLZQITUOI-UHFFFAOYSA-N |
Synonym | 4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole |
PubChem CID | 600848 |
IUPAC Name | 4-(trifluoromethoxy)aniline |
SMILES | C1=CC(=CC=C1N)OC(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10165650
|
Thermo Scientific Acros
297640025 |
2.5 g | Glass bottle |
27.50 €
2.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10582651
|
Thermo Scientific Acros
297640250 |
25 g | Glass bottle |
117.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
461-82-5 | |
177.12 | |
XUJFOSLZQITUOI-UHFFFAOYSA-N | |
600848 | |
C1=CC(=CC=C1N)OC(F)(F)F |
C7H6F3NO | |
MFCD00041314 | |
4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole | |
4-(trifluoromethoxy)aniline |
Specifications
461-82-5 | |
100.0 | |
1.3100g/mL | |
80°C | |
98.5% min. (GC) | |
CF3OC6H4NH2 | |
MFCD00041314 | |
4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole | |
C1=CC(=CC=C1N)OC(F)(F)F | |
177.12 | |
177.12 | |
Liquid |
99.0 | |
Colorless to Yellow | |
83.0°C to 85.0°C (15.0 mmHg) | |
Authentic | |
C7H6F3NO | |
1.4620 to 1.4640 | |
1.31 | |
XUJFOSLZQITUOI-UHFFFAOYSA-N | |
4-(trifluoromethoxy)aniline | |
600848 | |
99% | |
4-(Trifluoromethoxy)aniline, 99+% |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Fatal in contact with skin.
May cause damage to organs through prolonged or repeated exposure.<b
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 207-317-5
- 4-(Trifluoromethoxy)aniline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
RUO – Research Use Only