missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethyl)piperidine, 97%, Thermo Scientific Chemicals

60.40 € - 720.00 €

Chemical Identifiers

CAS 657-36-3
Molecular Formula C6H10F3N
Molecular Weight (g/mol) 153.15
MDL Number MFCD00102068
InChI Key RDRQUUWCJTYHCT-UHFFFAOYSA-N
Synonym 4-trifluoromethyl piperidine, 4-trifluoromethyl-piperidine, piperidine, 4-trifluoromethyl, pubchem22645, 4-trifluormethyl-piperidine
PubChem CID 136469
IUPAC Name 4-(trifluoromethyl)piperidine
SMILES FC(F)(F)C1CCNCC1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15452229
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H64840.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15432229
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H64840.03
1 g
182.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
15442229
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H64840.06
5 g
720.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

657-36-3
153.15
RDRQUUWCJTYHCT-UHFFFAOYSA-N
136469
FC(F)(F)C1CCNCC1
C6H10F3N
MFCD00102068
4-trifluoromethyl piperidine, 4-trifluoromethyl-piperidine, piperidine, 4-trifluoromethyl, pubchem22645, 4-trifluormethyl-piperidine
4-(trifluoromethyl)piperidine

Specifications

657-36-3
C6H10F3N
4-trifluoromethyl piperidine, 4-trifluoromethyl-piperidine, piperidine, 4-trifluoromethyl, pubchem22645, 4-trifluormethyl-piperidine
FC(F)(F)C1CCNCC1
153.15
153.15
Liquid
Colorless
MFCD00102068
RDRQUUWCJTYHCT-UHFFFAOYSA-N
4-(trifluoromethyl)piperidine
136469
97%
4-(Trifluoromethyl)piperidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only