missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Trifluoromethylsulfony)phenylacetic acid, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS | 1099597-82-6 |
Packaging | Amber Glass Bottle |
Molecular Formula | C9H7F3O4S |
Quantity | 1 g |
Synonym | 4-trifluoromethylsulfony phenylacetic acid, 2-4-trifluoromethyl sulfonyl phenyl acetic acid, 4-trifluoromethanesulfonylphenyl acetic acid, 4-trifluoromethanesulfonyl phenyl acetic acid, 4-trifluoromethylsulfonyl phenylacetic acid, 2-4-trifluoromethylsulfonyl phenyl acetic acid |
InChI Key | FIEUCHICQSJBAU-UHFFFAOYSA-N |
Molecular Weight (g/mol) | 268.21 |
PubChem CID | 40427164 |
Formula Weight | 268.21 |
Percent Purity | 97% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'