missing translation for 'onlineSavingsMsg'
Learn More

4-(Trifluoromethylsulfonyl)benzonitrile, 97+%, Thermo Scientific Chemicals

Product Code. 11449957
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312-21-0
Molecular Formula C8H4F3NO2S
Molecular Weight (g/mol) 235.18
MDL Number MFCD01631631
InChI Key UHCGSMMNICMDIX-UHFFFAOYSA-N
Synonym 4-trifluoromethylsulfonyl benzonitrile, 4-trifluoromethyl sulfonyl benzonitrile, 4-trifluoromethanesulfonyl benzonitrile, 4-trifluoromethanesulfonylbenzonitrile, 4-trifluoromethyl sulfonyl benzenecarbonitrile, acmc-1clj3, 4-trifluoromethanesulphonylbenzonitrile, 4-trifluoromethylsulfonyl benzonitril
PubChem CID 2777824
IUPAC Name 4-(trifluoromethylsulfonyl)benzonitrile
SMILES C1=CC(=CC=C1C#N)S(=O)(=O)C(F)(F)F

Specifications

Melting Point 87°C to 92°C
Quantity 5 g
UN Number UN3439
Formula Weight 235.18
Percent Purity ≥97%
Chemical Name or Material 4-(Trifluoromethylsulfonyl)benzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'