Learn More
Masterflex™ 4-way Stopcock
Use these stopcocks for a variety of applications. Cole-Parmer™ 4-way Stopcocks feature a PC body and an HDPE stem.
Brand:  Masterflex™ 30600-04
50.23 EUR valid until 2025-01-17
Use promo code "21645" to get your promotional price.
 View more versions of this product
Product Code. 11742683
  • 148.00 € / Pack of 10
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
Use these stopcocks for a variety of applications. Cole-Parmer™ 4-way Stopcocks feature a PC body and an HDPE stem.
  • Four-way stopcock
  • Choose stopcocks with either a male luer lock or a male luer slip
  • Handle rotates 360 degrees on four-way stopcocks
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Specifications
Specifications
Stopcock
Polycarbonate and HDPE
Male Luer Lock
Polycarbonate, HDPE
Male Luer Lock or A Male Luer Slip