missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ 40 L Selective Sorting Containers with Lids

40 L Selective Sorting Containers with Lids are adapted to selective sorting.

Brand:  Gilac™ G144800

Product Code. 17958731

  • 210.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Containers adapted to selective sorting.
  • Easily accessible.When stacked, the upper container allows the lid to be locked in the open position for ease of use.
  • Sold by pack of 3 containers with a blue lid, a yellow lid and a green lid for easy sorting of waste.
  • Stackable (with lid) and nestable (without lid) for storage.
  • Containers compatible with GILAC universal 600 x 400 rolling dollies for easy handling.
  • The dimensions indicated in the product features chart are the dimensions of a container with lid.
  • The height of 3 containers stacked: 1310 mm. The dimensions of the pack delivered in a box: 700 x 420 x 730 mm.
Specifications

Specifications

Storage Tote
Assorted
10.6 gal.
3 Pk.
HDPE
Rectangular
40 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilac™ 40 L Selective Sorting Containers with Lids > 40 L Selective Sorting Containers with Lids; Quantity: 3 Pk.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.