missing translation for 'onlineSavingsMsg'
Learn More

Stomacher™ 400 Circulator Lab Blender

A laboratory blender for easy and effective sample preparation, while delivering reliable and consistent results.

Brand:  Stomacher™ 0400/001/EU/W

Product Code. 17764389

  • 4790.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Suitable for volume sample sizes from 80 to 400mL
  • Programmable process times and paddle speeds for consistent results
  • Easy to use – simple to set up, programmable operation with digital control panel
  • Easy to clean
  • Durable and reliable, designed for constant use
Specifications

Specifications

240 V
140 W
Power cable, quick start guide, satchet of BA6141 Standard, and circulator Stomacher bags
330 x 355 x 300 mm
80 to 400 mL
Window version, 240 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.