Learn More
TubeWriter 400 - Standard Rack Top Print for 1.5-2.0 mL Microcentrifuge and Screw Cap Tubes
Standard rack for directly printing on the top of 1.5 to 2.0 mL microcentrifuge and screw cap tubes
Brand:  TubeWriter PT-1500MC-EL-400
Product Code. 30067415
  • 1105.00 € / Each
Explore more special offers
Description
Description
  • Standard printing rack designed for the 400
  • Prints and labels directly on the top of 1.5 to 2.0 mL microcentrifuge and screw cap tubes
  • Rack ensures correct alignment and holds tubes securely
  • No time-consuming sticky labels or handwriting required
  • Printing can take as little time as 50 tubes in 2 to 3 minutes
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Print withstands the harshest lab conditions
Specifications
Specifications
Labeling Rack