missing translation for 'onlineSavingsMsg'
Learn More

TubeWriter 400 - Standard Rack Top Print for 2.0 mL Cryogenic Vials

Standard rack for directly printing on the top of 2.0 mL cryogenic vials

Brand:  TubeWriter PT-1800CV-EL-400

Product Code. 30067442

  • 1105.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Standard printing rack designed for the 400
  • Prints and labels directly on the top of 2.0 mL cryogenic vials
  • Rack ensures correct alignment and holds vials securely
  • No time-consuming sticky labels or handwriting required
  • Printing can take as little time as 50 tubes in 2 to 3 minutes
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Print withstands the harshest lab conditions
Specifications

Specifications

Labeling Rack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers