missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ 406 Double Latex-Coated Insulated Cold-Weather Gloves

Product Code. 17737607
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
5 pairs
This item is not returnable. View return policy

Product Code. 17737607

Brand: SHOWA™ 40607M

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Water-repellant gloves are designed to remain dry when used with liquids, while intelligent acrylic insulation protects hands from freezing temperatures down to -30°C.

  • Double-dipped latex coating with a rough texture for enhanced grip capabilities
  • Intelligent acrylic insulation protects hands from temperatures down to -30°C
  • Insulation helps to reduce perspiration inside the glove to preserve grip and tactility
  • High visibility orange exterior increases user safety in low-light applications
  • Seamless knit construction for a snug fit to increase warmth and dexterity

Specifications

ANSI Abrasion Rating ANSI 2
Coated Area Palm
Construction Seamless Knit
Coating Material Latex
Color Hi-vis Orange, Black
Cut Resistance Rating ANSI A1
Exterior Finish Partially Textured
Hand Specific
Latex Free No
Material Nylon, Polyester
Size Medium
For Use With (Application) Brick Laying, Outdoor Maintenance, Refrigeration, and Shipping/Receiving
Glove Cuff Color Yellow
Glove Cuff Style Elasticated
Liner Material Acrylic
Puncture Rating ANSI/ISEA 105-16 Level 1
Series 406
Thumb Style Straight
Length (English) 11 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.