missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 43.2mm Septum for SCHOTT Screw Caps

Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.

Brand:  Fisherbrand™ 43 02 2095

 View more versions of this product

Product Code. 11510615

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • For use with SCHOTT Screw Caps
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Septa
43.2 mm
500
Gray, Red
Butyl, PTFE
2.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.