Learn More
Hamilton™ 1000 Series Gastight™ Saltline Syringes
For use with solutions that have a high salt concentration
Brand: Hamilton™ 208337
This item is not returnable.
View return policy
Description
- Equipped with inert and non-corrosive plunger
- Fitted with resistant ultra-high molecular weight poly-ethylene (UHMWPE) plunger tip
Applications:
For life science applications with salty buffers, mining applications with acids or trace metal analysis
Specifications
Not Autoclavable | |
Gastight Saltline Syringe | |
10 mL | |
Bubble Free Prime (BFP) |