Learn More
Hamilton™ 1000 Series Gastight™ Saltline Syringes
For use with solutions that have a high salt concentration
Brand:  Hamilton™ 208335
 View more versions of this product
Product Code. 15182514
  • 210.00 € / Each
Explore more special offers
Description
Description
  • Equipped with inert and non-corrosive plunger
  • Fitted with resistant ultra-high molecular weight poly-ethylene (UHMWPE) plunger tip
Applications:
For life science applications with salty buffers, mining applications with acids or trace metal analysis
Specifications
Specifications
Not Autoclavable
Gastight Saltline Syringe
1 mL
Bubble Free Prime (BFP)