Learn More
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Brand:  Hamilton™ 7638-01
 View more versions of this product
Product Code. 10079742
  • 80.00 € / Each
Explore more special offers
Description
Description
  • ideal for dispensing volumes from 0.05 µL up to 5 µL
  • The 7000 series is the only zero dead volume syringe
  • The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications
Specifications
710RN
22 G
710