Learn More
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included
Brand:  Hamilton™ 7658-01
 View more versions of this product
Product Code. 10335772
  • 84.00 € / Each
Explore more special offers
Description
Description
  • PTFE coating from point for 0.75 in.
  • For dispensing both liquids and gases
  • Precision-machined polymer plunger tip that creates leak-free seal
Available as replacement needle for installing into standard removable needle (RN) syringe; Needles sold separately
Specifications
Specifications
22 G
TLC
1750
500 μL