missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included

Brand:  Hamilton™ 7634-01

 View more versions of this product

Product Code. 10444805

  • 77.60 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • ideal for dispensing volumes from 0.05 µL up to 5 µL
  • The 7000 series is the only zero dead volume syringe
  • The fine gauge plunger wire travels to the tip of the needle ensuring that all solvent is expelled from the needle
Specifications

Specifications

Autoclavable
0.014 in.
0.343 mm
Sample spotting
2000 psi
115°C
75RN
10°C
5 μL
Removable
Borosilicate Glass (Barrel), Stainless Steel (Plunger)
0.260 in.
6.60 mm
Microliter Syringe
239°F
50°F
No Needle Included
Ea.
700 Series Microliter Syringes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included > Model 75 RN; Microliter; 5uL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.