Learn More
BRAND™ BRANDplates™ Insert System Microplates - 6-Well Specialized Plate
6-well specialized plate
Brand: BRAND™ 782891
91.19 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.58800kg
Description
- Transparent
- Available with pureGrade™ S and cellGrade™ plus surface
- Four wells connected as one large, elongated well, fitted with strip of 4 inserts
- Guide ridges for perfect insert fit
- Designed feeding ports for easy media changes, suitable for robotics/automation
- 10 plates/Pk. (Individually wrapped with lid)
Applications:
3D tissue culture
Compliance
ANSI/SLAS standards 1 and 4
Specifications
Sterile | |
Cell Culture Insert System | |
3D tissue culture |
cellGrade plus | |
6 | |
Insert system |