Learn More
Hamilton™ Microlab™ 1700 Series Gastight™ Syringes
For dispensing both liquids and gases
Brand:  Hamilton™ 80262
 View more versions of this product
Product Code. 11542385
  • 130.00 € / Each
Explore more special offers
Description
Description
  • Equipped with precision-machined polymer plunger tip, often PTFE, which creates leak-free seal
  • Tight fit feature ensures tip wipes interior of syringe barrel free of sample
  • Reduces the chance deposit will occur in samples with dissolved or suspended molecules and result in damaged plunger
  • ChemSeal termination
Specifications
Specifications
22s G
1702
25 μL