missing translation for 'onlineSavingsMsg'
Learn More

KleenGuard™ G35 Seamless Knit Nylon Inspection Gloves

Product Code. 15480472
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 10
Pack of 240
This item is not returnable. View return policy

Product Code. 15480472

Brand: KleenGuard™ 38719

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Low-lint gloves are ideal for detecting imperfections and defects when conducting critical quality control inspections. Ambidextrous KleenGuard™ G35 Seamless Knit Nylon Inspection Gloves are silicone-free for use in a wide variety of applications including film handling, aerospace, and automotive assembly.

  • Ambidextrous design allows gloves to be replaced one at a time, rather than in pairs
  • High degree of sensitivity and dexterity for inspecting for defects and imperfections
  • Designed for critical quality control inspections
  • Seamless knit nylon shell reduces skin irritation
  • Low lint for cleanroom applications
  • Silicone-free

Specifications

Construction Seamless Knit
Coating Material Uncoated
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nylon
Product Type Inspection Gloves
Size Large
Sterility Non-sterile
For Use With (Application) Automotive Assembly, Aerospace Engineering, and Quality Control Inspections
Glove Cuff Style Knit
Series KleenGuard™ G35
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.