missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble Chase™ 5.1 Borosilicate Glass Disposable Test Tubes
Measure, mix and pour using 5.1 Borosilicate Glass Disposable Test Tubes. Tubes are rimmed and are available in various lengths.
Brand: DWK Life Sciences A507008000521
This item is not returnable.
View return policy
Description
- Disposable
- Clear borosilicate glass
- Round bottom
- Rimmed
Specifications
Round Bottom | |
70 mm | |
100 Pack | |
Autoclavable |
Test tube | |
Borosilicate Glass | |
Yes | |
8 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction