Learn More
5-(2,5-Dichlorophenyl)furan-2-carbonyl chloride, 99%, Thermo Scientific™
82.00 € - 366.00 €
Chemical Identifiers
CAS | 380906-80-9 |
---|---|
Molecular Formula | C11H5Cl3O2 |
Molecular Weight (g/mol) | 275.509 |
MDL Number | MFCD02258039 |
InChI Key | BSRRSYCAENNRPG-UHFFFAOYSA-N |
Synonym | 5-2,5-dichlorophenyl furan-2-carbonyl chloride, 5-2,5-dichloro-phenyl-furan-2-carbonyl chloride |
PubChem CID | 2771971 |
IUPAC Name | 5-(2,5-dichlorophenyl)furan-2-carbonyl chloride |
SMILES | C1=CC(=C(C=C1Cl)C2=CC=C(O2)C(=O)Cl)Cl |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
380906-80-9 | |
275.509 | |
BSRRSYCAENNRPG-UHFFFAOYSA-N | |
2771971 | |
C1=CC(=C(C=C1Cl)C2=CC=C(O2)C(=O)Cl)Cl |
C11H5Cl3O2 | |
MFCD02258039 | |
5-2,5-dichlorophenyl furan-2-carbonyl chloride, 5-2,5-dichloro-phenyl-furan-2-carbonyl chloride | |
5-(2,5-dichlorophenyl)furan-2-carbonyl chloride |
Specifications
380906-80-9 | |
MFCD02258039 | |
Moisture sensitive | |
BSRRSYCAENNRPG-UHFFFAOYSA-N | |
5-(2,5-dichlorophenyl)furan-2-carbonyl chloride | |
2771971 | |
99% |
C11H5Cl3O2 | |
UN3261 | |
5-2,5-dichlorophenyl furan-2-carbonyl chloride, 5-2,5-dichloro-phenyl-furan-2-carbonyl chloride | |
C1=CC(=C(C=C1Cl)C2=CC=C(O2)C(=O)Cl)Cl | |
275.509 | |
275.52 | |
5-(2,5-Dichlorophenyl)furan-2-carbonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
Your input is important to us. Please complete this form to provide feedback related to the content on this product.