missing translation for 'onlineSavingsMsg'
Learn More

5-(2,6-Dichlorophenyl)-1H-tetrazole, 97%, Thermo Scientific™

Product Code. 11304136
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50907-31-8
Molecular Formula C7H4Cl2N4
Molecular Weight (g/mol) 215.04
MDL Number MFCD00808036
InChI Key RLXQNTWJPTUTCL-UHFFFAOYSA-N
Synonym 5-2,6-dichlorophenyl-1h-tetrazole, 5-2,6-dichlorophenyl-2h-tetrazole, 5-2,6-dichlorophenyl-1h-1,2,3,4-tetrazole, 5-2,6-dichlorophenyl-1h-1,2,3,4-tetraazole, maybridge1_003804, acmc-20ama2, 5-2,6-dichlorophenyl-1h-tetraazole, 1h-tetrazole, 5-2,6-dichlorophenyl

Specifications

Melting Point 203°C to 209°C
Quantity 1 g
Beilstein 5268246
Formula Weight 215.04
Percent Purity 97%
Chemical Name or Material 5-(2,6-Dichlorophenyl)-1H-tetrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'